Learn More
Medchemexpress LLC HY-101384A 5mg Medchemexpress, Ro 10-5824 (dihydrochloride) CAS:189744-94-3 Purity:>98%
SDP
Catalog No. 501932105
Encompass_Preferred
Medchemexpress, HY-101384A 5mg Ro 10-5824 (dihydrochloride) CAS:189744-94-3 Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with K i of 5.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2105 Supplier Medchemexpress LLC Supplier No. HY101384A5MG
Medchemexpress, HY-101384A 5mg Ro 10-5824 (dihydrochloride) CAS:189744-94-3 Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with K i of 5.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.