Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro 10-5824 dihydrochloride | 189744-94-3 | 99.9% | 353.29 | C17H22Cl2N4 | 1 ML
SDP

Catalog No. 5000212460
Encompass_Preferred

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid, and is characterized for potency, purity, and identity for receptor pharmacology and in vitro studies.

  • Selective dopamine D4 receptor partial agonist with reported Ki = 5.2 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) or as solid quantities for custom dosing.
  • High purity (reported 99.86%), suitable for analytical and biological assays.
  • Molecular weight 353.29, chemical formula C17H22Cl2N4, CAS 189744-94-3.
  • Characterized with supporting documentation including datasheet, COA, LCMS, and SDS.
  • Intended for receptor pharmacology, signaling studies, and in vitro assay applications.

Catalog No. 50-002-12460 Supplier Medchemexpress LLC Supplier No. HY101384A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid, and is characterized for potency, purity, and identity for receptor pharmacology and in vitro studies.

  • Selective dopamine D4 receptor partial agonist with reported Ki = 5.2 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) or as solid quantities for custom dosing.
  • High purity (reported 99.86%), suitable for analytical and biological assays.
  • Molecular weight 353.29, chemical formula C17H22Cl2N4, CAS 189744-94-3.
  • Characterized with supporting documentation including datasheet, COA, LCMS, and SDS.
  • Intended for receptor pharmacology, signaling studies, and in vitro assay applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.