Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro 10-5824 dihydrochloride | 189744-94-3 | 99.9% | 353.29 g/mol | C17H22Cl2N4 | 25 MG
SDP

Catalog No. 5000212831
Encompass_Preferred

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist used in pharmacology and neuroscience research. It binds D4 receptors with high affinity and is supplied as a white to off-white solid for laboratory use.

  • High purity (99.9%) for research applications.
  • Selective D4 receptor partial agonist with Ki ≈ 5.2 nM.
  • Molecular formula C17H22Cl2N4 and molecular weight 353.29 g/mol.
  • Available in small milligram quantities for experimental studies.
  • White to off-white solid suitable for handling and formulation.

Catalog No. 50-002-12831 Supplier Medchemexpress LLC Supplier No. HY101384A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist used in pharmacology and neuroscience research. It binds D4 receptors with high affinity and is supplied as a white to off-white solid for laboratory use.

  • High purity (99.9%) for research applications.
  • Selective D4 receptor partial agonist with Ki ≈ 5.2 nM.
  • Molecular formula C17H22Cl2N4 and molecular weight 353.29 g/mol.
  • Available in small milligram quantities for experimental studies.
  • White to off-white solid suitable for handling and formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.