Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro 10-5824 dihydrochloride | 189744-94-3 | 99.9% | 353.29 g/mol | C17H22Cl2N4 | 50 MG
SDP

Catalog No. 5000211450
Encompass_Preferred

Ro 10-5824 dihydrochloride is a research chemical used in neuropharmacology as a selective dopamine D4 receptor partial agonist. Supplied as the dihydrochloride salt with characterized molecular properties and high reported purity, it is offered in solid and solution formats for laboratory use.

  • Selective dopamine D4 receptor partial agonist with Ki ≈ 5.2 nM.
  • High reported purity (99.86%).
  • Molecular weight 353.29 g/mol and formula C17H22Cl2N4.
  • Available as solid powder and DMSO solution in small research pack sizes.
  • Dihydrochloride salt form improves handling and solubility.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-11450 Supplier Medchemexpress LLC Supplier No. HY101384A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 10-5824 dihydrochloride is a research chemical used in neuropharmacology as a selective dopamine D4 receptor partial agonist. Supplied as the dihydrochloride salt with characterized molecular properties and high reported purity, it is offered in solid and solution formats for laboratory use.

  • Selective dopamine D4 receptor partial agonist with Ki ≈ 5.2 nM.
  • High reported purity (99.86%).
  • Molecular weight 353.29 g/mol and formula C17H22Cl2N4.
  • Available as solid powder and DMSO solution in small research pack sizes.
  • Dihydrochloride salt form improves handling and solubility.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.