Learn More
Cayman Chemical Ro 106-9920 10mg
SDP
Catalog No. 502972962
Encompass_Preferred
A small molecule inhibitor of NF-κB-dependent expression of TNF-α, IL-1β, and IL-6 (IC50 50 = 2.3 µM)
Catalog No. 50-297-2962 Supplier Cayman Chemical Supplier No. 1537310 MG
A small molecule inhibitor of NF-κB-dependent expression of TNF-α, IL-1β, and IL-6 (IC50 50 = 2.3 µM)