Learn More
Apexbio Technology LLC Ro 106-9920 62645-28-7 10mg
SDP
Catalog No. 5000088553
Encompass_Preferred
Ro 106-9920 (CAS 62645-28-7) is a small molecule inhibitor that selectively suppresses the activation of the NF-kappaB signaling pathway By interfering with NF-kappaB function Ro 106-9920 modulates transcriptional events involved in cellular inflammatory responses survival and tumor progression Aberrant NF-kappaB activity is frequently observed in various cancer cell types and is linked to enhanced proliferation invasion and resistance to apoptosis Ro 106-9920 is widely utilized in cellular and animal studies to investigate the roles of NF-kappaB in oncogenesis and related pathologies providing a valuable tool for elucidating NF-kappaB-mediated mechanisms and evaluating potential therapeutic strategies
Catalog No. 50-000-88553 Supplier Apexbio Technology LLC Supplier No. B685310
Ro 106-9920 (CAS 62645-28-7) is a small molecule inhibitor that selectively suppresses the activation of the NF-kappaB signaling pathway By interfering with NF-kappaB function Ro 106-9920 modulates transcriptional events involved in cellular inflammatory responses survival and tumor progression Aberrant NF-kappaB activity is frequently observed in various cancer cell types and is linked to enhanced proliferation invasion and resistance to apoptosis Ro 106-9920 is widely utilized in cellular and animal studies to investigate the roles of NF-kappaB in oncogenesis and related pathologies providing a valuable tool for elucidating NF-kappaB-mediated mechanisms and evaluating potential therapeutic strategies