Learn More
Abcam Ro 106-9920, NF-kappaB inhibitor, 50MG
Catalog No. 502705524
Encompass_Preferred
MW 245.26 Da, Purity >95%. NF-κB inhibitor. Anti-inflammatory agent. Selectively inhibits LPS- and TNF-α-induced IκBα ubiquitination (IC₅₀ = 3 μM). Inhibits TNF-α, IL-1β and IL-6 production. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5524 Supplier Abcam Supplier No. AB14465550MG
MW 245.26 Da, Purity >95%. NF-κB inhibitor. Anti-inflammatory agent. Selectively inhibits LPS- and TNF-α-induced IκBα ubiquitination (IC₅₀ = 3 μM). Inhibits TNF-α, IL-1β and IL-6 production. Shows anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement