Learn More
American Radiolabeled Chemicals Inc RO 15-4513 3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1819250UC
Encompass
RO 15-4513-7,9-3H 250uCi in ethanol, 1 mCi/ml, S.A 20-40 Ci/mmol, M.W. 326.3, Shipped in Dry Ice
Catalog No. 50-901-140
Only null left
Add to Cart
Request a Quote