Learn More
Enzo Life Sciences RO-20-1724 (100mg). CAS: 29925-17-5
SDP
Supplier:  Enzo Life Sciences BMLEI1170100
Encompass_Preferred
Selective, potent cAMP-specific phosphodiesterase inhibitor (PDE IV, IC50=2µM). Inhibits superoxide generation and platelet aggregation stimulated by arachidonic acid. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells. Alternative name: 4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone. Purity: ≥99% (TLC). Solubility: Soluble in 100% ethanol (40mg/ml) or DMSO (200 mg/ml); insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0559