Learn More
TARGETMOL CHEMICALS INC Ro 20-1724 25MG
SDP
Catalog No. 502878307
Encompass_Preferred
Also available in 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Ro 20-1724 (4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone) is a widely used inhibitor of cyclic nucleotide phosphodiesterase with IC50 of 2.0 uM and Ki of 3.1 uM. Purity 98%
Catalog No. 50-287-8307 Supplier TARGETMOL CHEMICALS INC Supplier No. T1970625MG
Also available in 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Ro 20-1724 (4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone) is a widely used inhibitor of cyclic nucleotide phosphodiesterase with IC50 of 2.0 uM and Ki of 3.1 uM. Purity 98%