Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro 20-1724 | 29925-17-5 | MFCD00077397 | 100.0% | 278.35 g/mol | C15H22N2O3 | 25 MG
SDP

Catalog No. 5000212240
Encompass_Preferred

Ro 20-1724 is a research-grade small molecule inhibitor selective for cAMP-specific phosphodiesterase 4 (PDE4). Supplied as a solid reagent for in vitro and in vivo studies, it includes documented solubility, storage, and recommended stock solution information to support biochemical and pharmacology experiments.

  • Selective inhibitor of cAMP-specific phosphodiesterase 4 (PDE4) with reported Ki of 1930 nM.
  • High purity suitable for research applications.
  • Solid form with recommended storage at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO ≥ 50 mg/mL; in vivo formulations reported to achieve ≥ 2.5 mg/mL.
  • Molecular weight 278.35 g/mol and formula C15H22N2O3 for compound identification.
  • Available in small laboratory quantities to support dose-ranging studies.

Catalog No. 50-002-12240 Supplier Medchemexpress LLC Supplier No. HY10092725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 20-1724 is a research-grade small molecule inhibitor selective for cAMP-specific phosphodiesterase 4 (PDE4). Supplied as a solid reagent for in vitro and in vivo studies, it includes documented solubility, storage, and recommended stock solution information to support biochemical and pharmacology experiments.

  • Selective inhibitor of cAMP-specific phosphodiesterase 4 (PDE4) with reported Ki of 1930 nM.
  • High purity suitable for research applications.
  • Solid form with recommended storage at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO ≥ 50 mg/mL; in vivo formulations reported to achieve ≥ 2.5 mg/mL.
  • Molecular weight 278.35 g/mol and formula C15H22N2O3 for compound identification.
  • Available in small laboratory quantities to support dose-ranging studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.