Learn More
Enzo Life Sciences RO-20-1724, (1g), CAS Number: 29925-17-5
SDP
Catalog No. NC9885017
Encompass_Preferred
PDE inhibitor. Selective, potent cAMP-specific phosphodiesterase inhibitor (PDE IV, IC50=2µM). Inhibits superoxide generation and platelet aggregation stimulated by arachidonic acid. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells. Alternative Name: 4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone. Formula: C15H22N2O3. MW: 278.3. CAS Number: 29925-17-5. Purity: ≥99% (TLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol (40mg/ml) or DMSO (200 mg/ml); insoluble in water. Use/Stability: Stable for at least 1 year after receipt when stored at +20°C. Solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: Ambient
Catalog No. NC9885017 Supplier Enzo Life Sciences Supplier No. BMLEI1171000
PDE inhibitor. Selective, potent cAMP-specific phosphodiesterase inhibitor (PDE IV, IC50=2µM). Inhibits superoxide generation and platelet aggregation stimulated by arachidonic acid. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells. Alternative Name: 4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone. Formula: C15H22N2O3. MW: 278.3. CAS Number: 29925-17-5. Purity: ≥99% (TLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol (40mg/ml) or DMSO (200 mg/ml); insoluble in water. Use/Stability: Stable for at least 1 year after receipt when stored at +20°C. Solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: Ambient