Learn More
Medchemexpress LLC Ro 25-6981 Maleate | 1312991-76-6 | 99.25% | 455.54 | 100 MG
SDP
Catalog No. 5000352435
Encompass_Preferred
Ro 25-6981 Maleate is a potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist. It demonstrates anticonvulsant and anti-parkinsonian activity and has potential for research into Parkinson's disease.
  • Potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist
  • Demonstrates anticonvulsant activity
  • Demonstrates anti-parkinsonian activity
  • Potential for Parkinson's disease research
  • Induces contraversive rotations in 6-OHDA-lesioned rats
  • Exhibits age- and activation-dependent anticonvulsant action
  • Shows analgesic effects on incision pain in rats
Catalog No. 50-003-52435 Supplier Medchemexpress LLC Supplier No. HY13993A100MG
Ro 25-6981 Maleate is a potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist. It demonstrates anticonvulsant and anti-parkinsonian activity and has potential for research into Parkinson's disease.
  • Potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist
  • Demonstrates anticonvulsant activity
  • Demonstrates anti-parkinsonian activity
  • Potential for Parkinson's disease research
  • Induces contraversive rotations in 6-OHDA-lesioned rats
  • Exhibits age- and activation-dependent anticonvulsant action
  • Shows analgesic effects on incision pain in rats