Learn More
Medchemexpress LLC Ro 25-6981 Maleate | 1312991-76-6 | 99.3% | 455.54 | 1 ML
SDP
Catalog No. 5000351229
Encompass_Preferred
Ro 25-6981 Maleate is a potent, selective, and activity-dependent antagonist of the NMDA receptor containing the NR2B subunit. It demonstrates anticonvulsant and anti-parkinsonian activity, making it a valuable compound for Parkinson's disease research.
  • Potent and selective NMDA receptor antagonist
  • Specific for NR2B subunit
  • Demonstrates anticonvulsant activity
  • Demonstrates anti-parkinsonian activity
  • Useful for Parkinson's disease research
Catalog No. 50-003-51229 Supplier Medchemexpress LLC Supplier No. HY13993A10MM/1ML
Ro 25-6981 Maleate is a potent, selective, and activity-dependent antagonist of the NMDA receptor containing the NR2B subunit. It demonstrates anticonvulsant and anti-parkinsonian activity, making it a valuable compound for Parkinson's disease research.
  • Potent and selective NMDA receptor antagonist
  • Specific for NR2B subunit
  • Demonstrates anticonvulsant activity
  • Demonstrates anti-parkinsonian activity
  • Useful for Parkinson's disease research