Learn More
Medchemexpress LLC Ro 25-6981 (Maleate) | 1312991-76-6 | 99.3% | 455.54 | 50 MG
SDP
Catalog No. 5000348869
Encompass_Preferred
Ro 25-6981 Maleate is a potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist. It demonstrates anticonvulsant and anti-parkinsonian activity and is considered to have potential for research into Parkinson's disease (PD).
  • Potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist
  • Shows anticonvulsant activity
  • Shows anti-parkinsonian activity
  • Potential for Parkinson's disease (PD) research
Catalog No. 50-003-48869 Supplier Medchemexpress LLC Supplier No. HY13993A50MG
Ro 25-6981 Maleate is a potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist. It demonstrates anticonvulsant and anti-parkinsonian activity and is considered to have potential for research into Parkinson's disease (PD).
  • Potent, selective, and activity-dependent NR2B subunit-specific NMDA receptor antagonist
  • Shows anticonvulsant activity
  • Shows anti-parkinsonian activity
  • Potential for Parkinson's disease (PD) research