Learn More
Medchemexpress LLC Ro 25-6981 (maleate) | 1312991-76-6 | MFCD06798375 | 99.3% | C26H33NO6 | 10MG
SDP
Catalog No. 5000260434
Encompass_Preferred
Ro 25-6981 maleate is a research-grade small molecule that acts as a potent, selective, and activity-dependent antagonist of NR2B (GluN2B)-containing NMDA receptors. It has been used in preclinical studies demonstrating anticonvulsant, anti-parkinsonian, and analgesic effects and is supplied as a white to off-white solid with high purity.
  • Potent, selective NR2B NMDA receptor antagonist.
  • Demonstrated anticonvulsant and anti-parkinsonian activity in vivo.
  • High purity suitable for research applications.
  • Supplied as a white to off-white solid for easy handling.
  • Stable when stored sealed and protected from moisture.
Catalog No. 50-002-60434 Supplier Medchemexpress LLC Supplier No. HY13993A10MG
Ro 25-6981 maleate is a research-grade small molecule that acts as a potent, selective, and activity-dependent antagonist of NR2B (GluN2B)-containing NMDA receptors. It has been used in preclinical studies demonstrating anticonvulsant, anti-parkinsonian, and analgesic effects and is supplied as a white to off-white solid with high purity.
  • Potent, selective NR2B NMDA receptor antagonist.
  • Demonstrated anticonvulsant and anti-parkinsonian activity in vivo.
  • High purity suitable for research applications.
  • Supplied as a white to off-white solid for easy handling.
  • Stable when stored sealed and protected from moisture.