Learn More
TARGETMOL CHEMICALS INC Ro 25-6981 Maleate 25MG
SDP
Catalog No. 502878393
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Ro 25-6981 Maleate is a potent and selective activity-dependent NMDA receptor blocker, specifically targeting the NR2B subunit (IC50 values are 0.009 uM for NR1C/NR2B and 52 uM for NR1C/NR2A receptor subunit combinations). Purity 99.42%
Catalog No. 50-287-8393 Supplier TARGETMOL CHEMICALS INC Supplier No. T12758L25MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Ro 25-6981 Maleate is a potent and selective activity-dependent NMDA receptor blocker, specifically targeting the NR2B subunit (IC50 values are 0.009 uM for NR1C/NR2B and 52 uM for NR1C/NR2A receptor subunit combinations). Purity 99.42%