Learn More
Enzo Life Sciences Ro 31-8220 (1mg). CAS: 138489-18-6
SDP
Supplier:  Enzo Life Sciences BMLEI2830001
Encompass_Preferred
Cell-permeable PKC inhibitor. Selective for PKC (IC50=10nM) over CaM kinase II (IC50=17µM) and PKA (IC50=0.9µM). Induces apoptosis in various cell lines. Alternative name: 1, 1. Purity: ≥98%. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0667