Learn More
Medchemexpress LLC Ro 31-9790 | 145337-55-9 | 99.4% | 315.41 g/mol | C15H29N3O4 | 1 ML
SDP
Catalog No. 5000245645
Encompass_Preferred
Ro 31-9790 is a synthetic metalloproteinase (MMP) inhibitor used in biochemical and cell-based research. It blocks L-selectin shedding and modulates cytokine shedding in immune cells, displaying submicromolar to micromolar potency across different cell types. The compound is provided as a concentrated solution for laboratory assays and requires appropriate storage and handling.
  • Inhibits L-selectin and TNF-α shedding in immune cells.
  • Potent activity with IC50 values as low as 0.3 μM.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • High purity suitable for research applications.
  • High DMSO solubility (50 mg/mL) with warming or sonication if needed.
  • Stable in solvent when stored at -80°C for long-term preservation.
Catalog No. 50-002-45645 Supplier Medchemexpress LLC Supplier No. HY10170310MM 1ML
Ro 31-9790 is a synthetic metalloproteinase (MMP) inhibitor used in biochemical and cell-based research. It blocks L-selectin shedding and modulates cytokine shedding in immune cells, displaying submicromolar to micromolar potency across different cell types. The compound is provided as a concentrated solution for laboratory assays and requires appropriate storage and handling.
  • Inhibits L-selectin and TNF-α shedding in immune cells.
  • Potent activity with IC50 values as low as 0.3 μM.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • High purity suitable for research applications.
  • High DMSO solubility (50 mg/mL) with warming or sonication if needed.
  • Stable in solvent when stored at -80°C for long-term preservation.