Learn More
Medchemexpress LLC Ro 31-9790 (GI4747) | 145337-55-9 | 99.4% | 315.41 g/mol | C15H29N3O4 | 25 MG
SDP
Catalog No. 5000246977
Encompass_Preferred
Ro 31-9790 is a synthetic metalloproteinase (MMP) inhibitor used in research to block proteolytic shedding of cell-surface molecules such as L-selectin and TNF-α. It demonstrates sub-micromolar to low-micromolar activity in human and mouse immune cells and is supplied as a high-purity solid for laboratory experiments.
  • Potent inhibition of L-selectin shedding in immune cells.
  • Active against TNF-α shedding in monocytes.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes including small-milligram quantities.
  • Provided with datasheet, SDS, and COA for quality traceability.
Catalog No. 50-002-46977 Supplier Medchemexpress LLC Supplier No. HY10170325MG
Ro 31-9790 is a synthetic metalloproteinase (MMP) inhibitor used in research to block proteolytic shedding of cell-surface molecules such as L-selectin and TNF-α. It demonstrates sub-micromolar to low-micromolar activity in human and mouse immune cells and is supplied as a high-purity solid for laboratory experiments.
  • Potent inhibition of L-selectin shedding in immune cells.
  • Active against TNF-α shedding in monocytes.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes including small-milligram quantities.
  • Provided with datasheet, SDS, and COA for quality traceability.