Learn More
Medchemexpress LLC HY-12529 10mg Medchemexpress, Ro-3306 CAS:872573-93-8 Purity:>98%
SDP
Catalog No. 502028890
Encompass_Preferred
Medchemexpress, HY-12529 10mg Ro-3306 CAS:872573-93-8 Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8890 Supplier Medchemexpress LLC Supplier No. HY1252910MG
Medchemexpress, HY-12529 10mg Ro-3306 CAS:872573-93-8 Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.