Learn More
Medchemexpress LLC HY-12529 100mg , Ro-3306 CAS:872573-93-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12529100MG
Encompass_Preferred
Medchemexpress, HY-12529 100mg Ro-3306 CAS:872573-93-8 Formula:C18H13N3OS2 Ki: 20 nM (CDK1), 35 nM (CDK1/cyclin B1), 340 nM (CDK2/cyclin E), 318 nM (PKCδ) Purity:98% Ro-3306 is a potent and selective inhibitor of CDK1 , with K i s of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2967