Learn More
AdipoGen RO-3306
SDP
Catalog No. 502562338
Encompass_Preferred
Chemical. CAS 872573-93-8. Formula C18H13N3OS2. MW 351.5. Selective CDK 1 inhibitor Cdk1/B1 and Cdk1/A. Potential anti-cancer compound. Cell cycle arrest inducer at G1, S and G2/M . Apoptosis inducer in embryonic stem cells. Actively enhances downstream p53 signaling to promote apoptosis in AML cell lines. Suppresses phosphorylation of p53 at Ser315 and procaspase-8 at Ser387. Enhances Fas-mediated activation and processing of procaspase-8 in mitotic cells towards.
Catalog No. 50-256-2338 Supplier AdipoGen Supplier No. AGCR13515M025
Chemical. CAS 872573-93-8. Formula C18H13N3OS2. MW 351.5. Selective CDK 1 inhibitor Cdk1/B1 and Cdk1/A. Potential anti-cancer compound. Cell cycle arrest inducer at G1, S and G2/M . Apoptosis inducer in embryonic stem cells. Actively enhances downstream p53 signaling to promote apoptosis in AML cell lines. Suppresses phosphorylation of p53 at Ser315 and procaspase-8 at Ser387. Enhances Fas-mediated activation and processing of procaspase-8 in mitotic cells towards.