Learn More
Selleck Chemical LLC Ro-3306 10mg 872573-93-8
SDP
Catalog No. NC1193372
Encompass_Preferred
RO-3306 is an ATP-competitive, and selective CDK1 inhibitor with Ki of 20 nM, >15-fold selectivity against a diverse panel of human kinases. RO-3306 enhances p53-mediated Bax activation and mitochondrial apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1193372 Supplier Selleck Chemical LLC Supplier No. S774710MG
RO-3306 is an ATP-competitive, and selective CDK1 inhibitor with Ki of 20 nM, >15-fold selectivity against a diverse panel of human kinases. RO-3306 enhances p53-mediated Bax activation and mitochondrial apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.