Learn More
Medchemexpress LLC HY-100027A 5mg Medchemexpress, Ro 41-1049 (hydrochloride) CAS:127917-66-2 Purity:>98%
SDP
Catalog No. 501931698
Encompass_Preferred
Medchemexpress, HY-100027A 5mg Ro 41-1049 (hydrochloride) CAS:127917-66-2 Ro 41-1049 hydrochloride is a reversible and selective inhibitor of monoamine oxidase-A (MAO-A) . An homogeneous population of high affinity binding sites for [ 3 H]Ro 41-1049 is found in membrane preparations from human frontal cortex and placenta ( K d values of 16.5 and 64.4 nM, respectively) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1698 Supplier Medchemexpress LLC Supplier No. HY100027A5MG
Medchemexpress, HY-100027A 5mg Ro 41-1049 (hydrochloride) CAS:127917-66-2 Ro 41-1049 hydrochloride is a reversible and selective inhibitor of monoamine oxidase-A (MAO-A) . An homogeneous population of high affinity binding sites for [ 3 H]Ro 41-1049 is found in membrane preparations from human frontal cortex and placenta ( K d values of 16.5 and 64.4 nM, respectively) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.