Learn More
Medchemexpress LLC Ro 41-1049 hydrochloride | 127917-66-2 | MFCD00900596 | 100.0% | 301.77 g·mol⁻¹ | C12H13ClFN3OS | 1 ML
SDP
Catalog No. 5000215470
Encompass_Preferred
Ro 41-1049 hydrochloride is a reversible, selective monoamine oxidase A (MAO-A) inhibitor supplied for research use. It is available as a solid and as a pre-made 10 mM solution in DMSO (1 mL), and is provided with a high reported purity to support reproducible in vitro experiments.
  • Reversible, selective MAO-A inhibition suitable for biochemical assays.
  • Supplied as solid or pre-made 10 mM solution in DMSO (1 mL) for convenience.
  • High reported purity to reduce confounding impurities.
  • Characterized by CAS number 127917-66-2 and known molecular weight for accurate dosing.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-15470 Supplier Medchemexpress LLC Supplier No. HY100027A10MM 1ML
Ro 41-1049 hydrochloride is a reversible, selective monoamine oxidase A (MAO-A) inhibitor supplied for research use. It is available as a solid and as a pre-made 10 mM solution in DMSO (1 mL), and is provided with a high reported purity to support reproducible in vitro experiments.
  • Reversible, selective MAO-A inhibition suitable for biochemical assays.
  • Supplied as solid or pre-made 10 mM solution in DMSO (1 mL) for convenience.
  • High reported purity to reduce confounding impurities.
  • Characterized by CAS number 127917-66-2 and known molecular weight for accurate dosing.
  • Intended for research use only; not for human or clinical use.