Learn More
Enzo Life Sciences Ro 48-8071 (50mg). CAS: 189197-69-1
SDP
Catalog No. 502010727
Encompass_Preferred
Inhibits cholesterol biosynthesis at the level of 2,3-oxidosqualene cyclase (OSC). Blocks OSC and cholesterol synthesis in HepG2 cells in the nanomolar range. A novel lipid lowering agent. Purity: ≥95% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0727 Supplier Enzo Life Sciences Supplier No. BMLEI3420050
Inhibits cholesterol biosynthesis at the level of 2,3-oxidosqualene cyclase (OSC). Blocks OSC and cholesterol synthesis in HepG2 cells in the nanomolar range. A novel lipid lowering agent. Purity: ≥95% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: Ambient.