Learn More
Medchemexpress LLC Ro 48-8071 Fumarate | 189197-69-1 | 99.13% | 564.44 | 5 MG
SDP
Catalog No. 5000388038
Encompass_Preferred
Ro 48-8071 is an inhibitor of oxidosqualene cyclase (OSC) with an IC50 of approximately 6.5 nM. It has demonstrated the ability to reduce cholesterol synthesis in HepG2 cells with an IC50 value of approximately 1.5 nM. In various cancer cell lines, Ro 48-8071 has shown cytotoxicity and can induce apoptosis. It has also been observed to lower LDL-C in hamsters and significantly reduce in vivo tumor growth in mice without causing weight loss.
  • Inhibits oxidosqualene cyclase.
  • Reduces cholesterol synthesis.
  • Exhibits cytotoxicity against certain cancer cells.
  • Induces apoptosis in specific cancer cell lines.
  • Lowers ldl-c in animal models.
  • Reduces in vivo tumor growth.
Catalog No. 50-003-88038 Supplier Medchemexpress LLC Supplier No. HY18630A5MG
Ro 48-8071 is an inhibitor of oxidosqualene cyclase (OSC) with an IC50 of approximately 6.5 nM. It has demonstrated the ability to reduce cholesterol synthesis in HepG2 cells with an IC50 value of approximately 1.5 nM. In various cancer cell lines, Ro 48-8071 has shown cytotoxicity and can induce apoptosis. It has also been observed to lower LDL-C in hamsters and significantly reduce in vivo tumor growth in mice without causing weight loss.
  • Inhibits oxidosqualene cyclase.
  • Reduces cholesterol synthesis.
  • Exhibits cytotoxicity against certain cancer cells.
  • Induces apoptosis in specific cancer cell lines.
  • Lowers ldl-c in animal models.
  • Reduces in vivo tumor growth.