Learn More
Medchemexpress LLC Ro 48-8071 fumarate | 189197-69-1 | MFCD05865242 | 99.1% | 564.44 g/mol | C27H31BrFNO6 | 10 MG
SDP
Catalog No. 5000294881
Encompass_Preferred
Ro 48-8071 fumarate is a potent oxidosqualene cyclase (OSC) inhibitor used in research to block cholesterol biosynthesis and to probe cholesterol-dependent cellular pathways in vitro and in vivo. It is supplied as the fumarate salt at high purity for biochemical and cell-based studies.
  • Potent oxidosqualene cyclase inhibitor (IC50 ≈ 6.5 nM).
  • Suitable for cell-based and in vivo research applications.
  • High purity appropriate for biochemical assays (≈99.1%).
  • Provided as the fumarate salt to aid stability and handling.
  • Molecular weight 564.44 g/mol for accurate dosing and calculations.
  • Supplied in small research pack sizes for screening and mechanistic studies.
Catalog No. 50-002-94881 Supplier Medchemexpress LLC Supplier No. HY18630A10MG
Ro 48-8071 fumarate is a potent oxidosqualene cyclase (OSC) inhibitor used in research to block cholesterol biosynthesis and to probe cholesterol-dependent cellular pathways in vitro and in vivo. It is supplied as the fumarate salt at high purity for biochemical and cell-based studies.
  • Potent oxidosqualene cyclase inhibitor (IC50 ≈ 6.5 nM).
  • Suitable for cell-based and in vivo research applications.
  • High purity appropriate for biochemical assays (≈99.1%).
  • Provided as the fumarate salt to aid stability and handling.
  • Molecular weight 564.44 g/mol for accurate dosing and calculations.
  • Supplied in small research pack sizes for screening and mechanistic studies.