Learn More
TARGETMOL CHEMICALS INC Ro 48-8071 fumarate 25MG
SDP
Catalog No. 502878626
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ro 48-8071 fumarate, an inhibitor of OSC (Oxidosqualene cyclase; IC50=6.5 nM), exhibits LDL (low-density lipoprotein) cholesterol-lowering activity. Purity 99.32%
Catalog No. 50-287-8626 Supplier TARGETMOL CHEMICALS INC Supplier No. T177125MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ro 48-8071 fumarate, an inhibitor of OSC (Oxidosqualene cyclase; IC50=6.5 nM), exhibits LDL (low-density lipoprotein) cholesterol-lowering activity. Purity 99.32%