Learn More
Medchemexpress LLC HY-18652 10mg , Ro 5126766 CAS:946128-88-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1865210MG
Encompass_Preferred
HY-18652 10mg Ro 5126766 CAS: 946128-88-7 Ro 5126766 (CH5126766) is a first-in-class dual MEK/RAF inhibitor that allosterically inhibits BRAFV600E, CRAF, MEK, and BRAF (IC50: 8.2, 56, 160 nM, and 190 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1020