Learn More
TARGETMOL CHEMICALS INC RO 5126766 5MG
SDP
Catalog No. 502576400
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO5126766 (CH5126766) is a dual RAF/MEK inhibitor with IC50 of 8.2 nM 19 nM 56 nM and 160 nM for BRAF V600E BRAF CRAF and MEK1 respectively. Phase 1. purity: 98%
Catalog No. 50-257-6400 Supplier TARGETMOL CHEMICALS INC Supplier No. T69715MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO5126766 (CH5126766) is a dual RAF/MEK inhibitor with IC50 of 8.2 nM 19 nM 56 nM and 160 nM for BRAF V600E BRAF CRAF and MEK1 respectively. Phase 1. purity: 98%