Learn More
Medchemexpress LLC RO-5963 | 1416663-77-8 | 98.2% | 100 MG
SDP
Catalog No. 5000233490
Encompass_Preferred
RO-5963 is a small-molecule dual inhibitor of p53 interactions with MDM2 and MDMX, reported with biochemical IC50s of approximately 17 nM (MDM2) and 24 nM (MDMX). It is intended for research use and is characterized by a defined molecular formula and high chemical purity.
  • Dual inhibitor of p53-MDM2 and p53-MDMX.
  • Biochemical IC50s of ~17 nM (MDM2) and ~24 nM (MDMX).
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90.
  • High purity suitable for research (98.2%).
  • Available in multiple package sizes including 100 MG.
Catalog No. 50-002-33490 Supplier Medchemexpress LLC Supplier No. HY120086100MG
RO-5963 is a small-molecule dual inhibitor of p53 interactions with MDM2 and MDMX, reported with biochemical IC50s of approximately 17 nM (MDM2) and 24 nM (MDMX). It is intended for research use and is characterized by a defined molecular formula and high chemical purity.
  • Dual inhibitor of p53-MDM2 and p53-MDMX.
  • Biochemical IC50s of ~17 nM (MDM2) and ~24 nM (MDMX).
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90.
  • High purity suitable for research (98.2%).
  • Available in multiple package sizes including 100 MG.