Learn More
Medchemexpress LLC RO-5963 | 1416663-77-8 | 98.2% | 25 MG
SDP
Catalog No. 5000231862
Encompass_Preferred
RO-5963 is a small-molecule dual inhibitor of p53-MDM2 and p53-MDMX, reported to have low-nanomolar activity (IC50 ≈ 17 nM for MDM2, ≈ 24 nM for MDMX). It is supplied as a research reagent for laboratory biochemical and cell-based studies.
  • Dual inhibitor of p53-MDM2 and p53-MDMX with low-nanomolar potency.
  • Suitable for biochemical and cell-based assays probing p53 pathway.
  • Reported purity 98.2% and characterized molecular properties.
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90 g·mol⁻1.
  • Available in small research pack sizes and recommended cold storage for stability.
Catalog No. 50-002-31862 Supplier Medchemexpress LLC Supplier No. HY12008625MG
RO-5963 is a small-molecule dual inhibitor of p53-MDM2 and p53-MDMX, reported to have low-nanomolar activity (IC50 ≈ 17 nM for MDM2, ≈ 24 nM for MDMX). It is supplied as a research reagent for laboratory biochemical and cell-based studies.
  • Dual inhibitor of p53-MDM2 and p53-MDMX with low-nanomolar potency.
  • Suitable for biochemical and cell-based assays probing p53 pathway.
  • Reported purity 98.2% and characterized molecular properties.
  • Molecular formula C24H21ClF2N4O5; molecular weight 518.90 g·mol⁻1.
  • Available in small research pack sizes and recommended cold storage for stability.