Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro-5963 | 1416663-77-8 | 98.2% | 50 MG
SDP

Catalog No. 5000231839
Encompass_Preferred

RO-5963 is a small-molecule dual inhibitor of p53-MDM2 and p53-MDMX that activates the p53 pathway and induces apoptosis in cancer cell lines. It exhibits nanomolar potency and has demonstrated increased apoptotic activity compared with Nutlin in MCF7 and ZR75-30 cells.

  • Dual p53-MDM2 and p53-MDMX inhibition (IC50 ≈ 17 nM and 24 nM)
  • High in vitro potency and apoptosis induction in breast cancer cell lines
  • Soluble in DMSO at 150 mg/mL for in vitro use
  • Available as solids in multiple milligram sizes and as DMSO solutions
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)

Catalog No. 50-002-31839 Supplier Medchemexpress LLC Supplier No. HY12008650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO-5963 is a small-molecule dual inhibitor of p53-MDM2 and p53-MDMX that activates the p53 pathway and induces apoptosis in cancer cell lines. It exhibits nanomolar potency and has demonstrated increased apoptotic activity compared with Nutlin in MCF7 and ZR75-30 cells.

  • Dual p53-MDM2 and p53-MDMX inhibition (IC50 ≈ 17 nM and 24 nM)
  • High in vitro potency and apoptosis induction in breast cancer cell lines
  • Soluble in DMSO at 150 mg/mL for in vitro use
  • Available as solids in multiple milligram sizes and as DMSO solutions
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.