Learn More
TARGETMOL CHEMICALS INC RO-5963 5MG
SDP
Catalog No. 502581155
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO-5963 is a dual inhibitor of p53-MDM2 and p53-MDMX (IC50s ~17 nM and ~24 nM respectively). purity: 98%
Catalog No. 50-258-1155 Supplier TARGETMOL CHEMICALS INC Supplier No. T167715MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO-5963 is a dual inhibitor of p53-MDM2 and p53-MDMX (IC50s ~17 nM and ~24 nM respectively). purity: 98%