Learn More
Apexbio Technology LLC Ro 60-0175 fumarate 169675-09-6 10mg
SDP
Catalog No. 5000091844
Encompass_Preferred
Ro 60-0175 fumarate (CAS 169675-09-6) is a selective agonist of the 5-HT2C serotonin receptor known to specifically activate serotonergic signaling pathways By targeting 5-HT2C receptors it modulates neurotransmitter release and neuronal circuitry influencing processes such as appetite regulation mood stabilization and cognitive function Ro 60-0175 fumarate is widely utilized in neuropharmacological research to investigate central nervous system mechanisms and to assess the pharmacological properties and therapeutic potential of 5-HT2C receptor agonists in models of depression anxiety obesity and related neuropsychiatric conditions
Catalog No. 50-000-91844 Supplier Apexbio Technology LLC Supplier No. B687810
Ro 60-0175 fumarate (CAS 169675-09-6) is a selective agonist of the 5-HT2C serotonin receptor known to specifically activate serotonergic signaling pathways By targeting 5-HT2C receptors it modulates neurotransmitter release and neuronal circuitry influencing processes such as appetite regulation mood stabilization and cognitive function Ro 60-0175 fumarate is widely utilized in neuropharmacological research to investigate central nervous system mechanisms and to assess the pharmacological properties and therapeutic potential of 5-HT2C receptor agonists in models of depression anxiety obesity and related neuropsychiatric conditions