Learn More
Apexbio Technology LLC Ro 61-8048 199666-03-0 10mg
SDP
Catalog No. 5000089828
Encompass_Preferred
Ro 61-8048 is a small-molecule inhibitor targeting kynurenine 3-hydroxylase (KMO) It is designed to selectively inhibit the catalytic conversion of kynurenine into 3-hydroxykynurenine thereby modulating the kynurenine pathway of tryptophan metabolism Ro 61-8048 exerts its biological activity primarily through selective KMO inhibition In in vitro studies Ro 61-8048 demonstrates inhibitory potency with an IC50 value of approximately 37 nM Based on these pharmacological properties Ro 61-8048 holds research potential in investigations of neurodegenerative and central nervous system immune-related disorders
Catalog No. 50-000-89828 Supplier Apexbio Technology LLC Supplier No. B733410
Ro 61-8048 is a small-molecule inhibitor targeting kynurenine 3-hydroxylase (KMO) It is designed to selectively inhibit the catalytic conversion of kynurenine into 3-hydroxykynurenine thereby modulating the kynurenine pathway of tryptophan metabolism Ro 61-8048 exerts its biological activity primarily through selective KMO inhibition In in vitro studies Ro 61-8048 demonstrates inhibitory potency with an IC50 value of approximately 37 nM Based on these pharmacological properties Ro 61-8048 holds research potential in investigations of neurodegenerative and central nervous system immune-related disorders