Learn More
Medchemexpress LLC Ro 61-8048 | 199666-03-0 | MFCD11040807 | 99.8% | 421.45 | C17H15N3O6S2 | 5MG
SDP
Catalog No. 5000238456
Encompass_Preferred
Ro 61-8048 is a small-molecule inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and pharmacological research to modulate the kynurenine pathway and study neurological disease models. The compound is provided in solid form and as a 10 mM solution in DMSO; formula C17H15N3O6S2, molecular weight 421.45 g/mol.
  • Potent KMO inhibitor with reported nanomolar activity.
  • High purity (99.76%) as specified by the supplier.
  • Available as solid (5 mg-200 mg) and as 10 mM solution in DMSO.
  • Well characterized: C17H15N3O6S2, 421.45 g/mol.
  • Suitable for in vitro and in vivo studies of the kynurenine pathway and neurological disease models.
Catalog No. 50-002-38456 Supplier Medchemexpress LLC Supplier No. HY123475MG
Add to Cart
Add to Cart
Ro 61-8048 is a small-molecule inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and pharmacological research to modulate the kynurenine pathway and study neurological disease models. The compound is provided in solid form and as a 10 mM solution in DMSO; formula C17H15N3O6S2, molecular weight 421.45 g/mol.
  • Potent KMO inhibitor with reported nanomolar activity.
  • High purity (99.76%) as specified by the supplier.
  • Available as solid (5 mg-200 mg) and as 10 mM solution in DMSO.
  • Well characterized: C17H15N3O6S2, 421.45 g/mol.
  • Suitable for in vitro and in vivo studies of the kynurenine pathway and neurological disease models.