Learn More
Medchemexpress LLC Ro 61-8048 | 199666-03-0 | MFCD11040807 | 99.8% | 421.45 g·mol⁻¹ | C17H15N3O6S2 | 10MG
SDP
Catalog No. 5000238724
Encompass_Preferred
Ro 61-8048 is an orally active, selective inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and pharmacological research. It has a reported IC50 of 37 nM and has been shown to increase extracellular kynurenic acid concentrations. The compound is provided as a high-purity solid and in solution formats, with supplier-provided storage recommendations, SDS, and batch certificates available.
Catalog No. 50-002-38724 Supplier Medchemexpress LLC Supplier No. HY1234710MG
Ro 61-8048 is an orally active, selective inhibitor of kynurenine 3-hydroxylase (KMO) used in biochemical and pharmacological research. It has a reported IC50 of 37 nM and has been shown to increase extracellular kynurenic acid concentrations. The compound is provided as a high-purity solid and in solution formats, with supplier-provided storage recommendations, SDS, and batch certificates available.