Learn More
Medchemexpress LLC HY-12844 1mg Medchemexpress, Ro 64-6198 CAS:280783-56-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128441MG
Encompass_Preferred
Medchemexpress, HY-12844 1mg Ro 64-6198 CAS:280783-56-4 Ro 64-6198 is a potent, selective, nonpeptide, high-affinity, high cellular permeability and brain penetration N/OFQ receptor (NOP) agonist with an EC50 value of 25.6 nM. Ro 64-6198 is at least 100 times more selective for the NOP receptor over the classic opioid receptors. Ro 64-6198 can be used for stress and anxiety, addiction, neuropathic pain, cough, and anorexia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9748