Learn More
Abcam Ro 67-4853, Positive allosteric modulator of mGlu1, 50MG
Catalog No. 502707068
Encompass_Preferred
MW 325.36 g/mol, Purity >99%. Potent, selective positive allosteric modulator for mGlu₁ (pEC₅₀ = 7.16 for rat mGlu₁a receptors). Increases basal mGluR₁-induced cAMP accumulation and potentiates glutamate-induced cAMP accumulation. Selectively enhanced responses to DHPG in vivo (EC₅₀= 95 nM in CA3 neurons).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7068 Supplier Abcam Supplier No. AB12097950MG
MW 325.36 g/mol, Purity >99%. Potent, selective positive allosteric modulator for mGlu₁ (pEC₅₀ = 7.16 for rat mGlu₁a receptors). Increases basal mGluR₁-induced cAMP accumulation and potentiates glutamate-induced cAMP accumulation. Selectively enhanced responses to DHPG in vivo (EC₅₀= 95 nM in CA3 neurons).
The product is subject to the following: Abcam Restricted Use Statement