Learn More
TARGETMOL CHEMICALS INC RO 67-7476 10MG
SDP
Catalog No. 502576936
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro 67-7476 is a positive allosteric modulator of mGlu1 receptors. No activity at human mGlu1 receptors. Potentiates glutamate-induced calcium release with EC 50 of 60.1 nM. purity: 99%
Catalog No. 50-257-6936 Supplier TARGETMOL CHEMICALS INC Supplier No. T347810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro 67-7476 is a positive allosteric modulator of mGlu1 receptors. No activity at human mGlu1 receptors. Potentiates glutamate-induced calcium release with EC 50 of 60.1 nM. purity: 99%