Learn More
Medchemexpress LLC Ro 90-7501 | 293762-45-5 | 99.1% | 340.38 g/mol | C20H16N6 | 100 MG
SDP
Catalog No. 5000247894
Encompass_Preferred
Ro 90-7501 is a small-molecule research compound that inhibits amyloid β42 (Aβ42) fibril assembly and reduces Aβ42-induced cytotoxicity. It is characterized by CAS 293762-45-5, molecular formula C20H16N6, molecular weight 340.38 g/mol, and reported purity of approximately 99.1%.
  • Inhibits amyloid β42 fibril assembly and reduces Aβ42-induced cytotoxicity (EC50 2 μM).
  • Well characterized: CAS 293762-45-5 and molecular weight 340.38 g/mol.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes including 100 mg for laboratory use.
Catalog No. 50-002-47894 Supplier Medchemexpress LLC Supplier No. HY103241100MG
Ro 90-7501 is a small-molecule research compound that inhibits amyloid β42 (Aβ42) fibril assembly and reduces Aβ42-induced cytotoxicity. It is characterized by CAS 293762-45-5, molecular formula C20H16N6, molecular weight 340.38 g/mol, and reported purity of approximately 99.1%.
  • Inhibits amyloid β42 fibril assembly and reduces Aβ42-induced cytotoxicity (EC50 2 μM).
  • Well characterized: CAS 293762-45-5 and molecular weight 340.38 g/mol.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes including 100 mg for laboratory use.