Learn More
Medchemexpress LLC HY-10870 5mg Medchemexpress, RO-9187 CAS:876708-03-1 Purity:>98%
SDP
Catalog No. 501968612
Encompass_Preferred
Medchemexpress, HY-10870 5mg RO-9187 CAS:876708-03-1 RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8612 Supplier Medchemexpress LLC Supplier No. HY108705MG
Medchemexpress, HY-10870 5mg RO-9187 CAS:876708-03-1 RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.