Learn More
Medchemexpress LLC HY-108912 5mg Medchemexpress, RO1138452 CAS:221529-58-4 Purity:>98%
SDP
Catalog No. 501968634
Encompass_Preferred
Medchemexpress, HY-108912 5mg RO1138452 CAS:221529-58-4 RO1138452 is a potent and selective IP (prostacyclin) receptor antagonist. RO1138452 displays high affinity for IP receptors. In human platelets, pKi is 9.3±0.1; in a recombinant IP receptor system, pKi is 8.7±0.06. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8634 Supplier Medchemexpress LLC Supplier No. HY1089125MG
Medchemexpress, HY-108912 5mg RO1138452 CAS:221529-58-4 RO1138452 is a potent and selective IP (prostacyclin) receptor antagonist. RO1138452 displays high affinity for IP receptors. In human platelets, pKi is 9.3±0.1; in a recombinant IP receptor system, pKi is 8.7±0.06. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.