Learn More
Medchemexpress LLC HY-103476 1mg Medchemexpress, Ro15-4513 CAS:91917-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034761MG
Encompass_Preferred
Medchemexpress, HY-103476 1mg Ro15-4513 CAS:91917-65-6 Ro15-4513, imidazobenzodiazepinone derivative, is a partial inverse agonist of benzodiazepine receptor (BZR). Ro15-4513 is a potent ethanol antagonist. Ro15-4513 has anti-anxiety effect[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8117