Learn More
TARGETMOL CHEMICALS INC RO24-7429 5MG
SDP
Catalog No. 502580504
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro24-7429 is an effective and orally active antagonist of HIV-1 transactivator protein Tat and an inhibitor of a runt-related transcription factor 1. Ro24-7429 has anti-HIV antifibrotic and anti-inflammatory effects. purity: 99%
Catalog No. 50-258-0504 Supplier TARGETMOL CHEMICALS INC Supplier No. T285785MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro24-7429 is an effective and orally active antagonist of HIV-1 transactivator protein Tat and an inhibitor of a runt-related transcription factor 1. Ro24-7429 has anti-HIV antifibrotic and anti-inflammatory effects. purity: 99%