Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro3201195 | 249937-52-8 | 371.36 | C19H18FN3O4 | 100 MG
SDP

Catalog No. 5000226650
Encompass_Preferred

RO3201195 is a research-grade small-molecule inhibitor of p38 mitogen-activated protein kinase (p38 MAPK). It inhibits WS cell proliferation with a reported IC50 of 190 nM. The compound has molecular formula C19H18FN3O4 and molecular weight 371.36 g·mol⁻¹ and is supplied for in vitro research use.

  • Inhibits p38 MAPK with reported cellular IC50 of 190 nM.
  • Molecular formula C19H18FN3O4; molecular weight 371.36 g·mol⁻¹.
  • Soluble in DMSO (50 mg/mL; may require ultrasonic treatment).
  • Powder storage: -20°C (stable for 3 years); in solvent: -80°C for 6 months or -20°C for 1 month.
  • Provided in small research pack sizes suitable for laboratory studies.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-26650 Supplier Medchemexpress LLC Supplier No. HY118612100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO3201195 is a research-grade small-molecule inhibitor of p38 mitogen-activated protein kinase (p38 MAPK). It inhibits WS cell proliferation with a reported IC50 of 190 nM. The compound has molecular formula C19H18FN3O4 and molecular weight 371.36 g·mol⁻¹ and is supplied for in vitro research use.

  • Inhibits p38 MAPK with reported cellular IC50 of 190 nM.
  • Molecular formula C19H18FN3O4; molecular weight 371.36 g·mol⁻¹.
  • Soluble in DMSO (50 mg/mL; may require ultrasonic treatment).
  • Powder storage: -20°C (stable for 3 years); in solvent: -80°C for 6 months or -20°C for 1 month.
  • Provided in small research pack sizes suitable for laboratory studies.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.