Learn More
Apexbio Technology LLC Ro3280 1062243-51-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062410
Encompass_Preferred
Ro3280 (CAS 1062243-51-9) is a selective small-molecule inhibitor targeting Polo-like kinase 1 (PLK1) exerting inhibitory activity with an IC50 of 3 nM PLK1 is a cell cycle kinase involved prominently in regulating progression from G2 to M phase frequently overexpressed in various malignancies Ro3280 inhibits PLK1 activity and induces apoptosis and cell cycle disruption in acute leukemia cell lines (e g U937 HL-60 NB4 K562 MV4-11) and diverse solid tumor models (e g lung carcinoma H82 colon carcinoma HT-29 breast carcinoma MDA-MB-468) In mouse xenografts of HT-29 tumors Ro3280 demonstrates significant tumor regression highlighting its potential in cancer therapeutics research
Catalog No. 50-000-62410 Supplier Apexbio Technology LLC Supplier No. A86815.1
Ro3280 (CAS 1062243-51-9) is a selective small-molecule inhibitor targeting Polo-like kinase 1 (PLK1) exerting inhibitory activity with an IC50 of 3 nM PLK1 is a cell cycle kinase involved prominently in regulating progression from G2 to M phase frequently overexpressed in various malignancies Ro3280 inhibits PLK1 activity and induces apoptosis and cell cycle disruption in acute leukemia cell lines (e g U937 HL-60 NB4 K562 MV4-11) and diverse solid tumor models (e g lung carcinoma H82 colon carcinoma HT-29 breast carcinoma MDA-MB-468) In mouse xenografts of HT-29 tumors Ro3280 demonstrates significant tumor regression highlighting its potential in cancer therapeutics research